Please login to your online account to display your discounted pricing

Barium Hydroxide, 8-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$585.30 - $585.30

Chemical Identifiers

Chemical Name or Material Barium Hydroxide
CAS 12230-71-6
Name Note 8-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Ba(OH)2·8H2O
Formula Weight 315.46
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002408

 
avantor performance
100605
Poly Bottle 2.5kg Each for $585.30
Description & Specifications

Specifications

Formula Weight 315.46
Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
CAS 12230-71-6
Chemical Name or Material Barium Hydroxide
Molecular Formula Ba(OH)2·8H2O
Packaging Poly Bottle
Specific Gravity 1L = 2.18kg
Quantity 2.5kg
Name Note 8-Hydrate