Please login to your online account to display your discounted pricing

Barium Hydroxide, 8-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  1006-01

 View all options for this product

Catalog No. 02002410

 

Description & Specifications

Chemical Identifiers

Packaging Poly Bottle
Quantity 500g
Specific Gravity 1L = 2.18kg
Quantity 500g
Chemical Name or Material Barium Hydroxide
Name Note 8-Hydrate
CAS 12230-71-6
Purity Grade BAKER ANALYZED™ ACS Reagent
Physical Form Crystal
Molecular Formula Ba(OH)2·8H2O
Formula Weight 315.46g/mol

Specifications

Chemical Name or Material Barium Hydroxide
CAS 12230-71-6
Name Note 8-Hydrate
Purity Grade BAKER ANALYZED™ ACS Reagent
Molecular Formula Ba(OH)2·8H2O
Formula Weight 315.46g/mol