Please login to your online account to display your discounted pricing

Barium hydrogen phosphate, White powder, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Barium hydrogen phosphate
CAS 10048-98-3
Physical Form Powder
Molecular Formula BaHPO4
Formula Weight 233.31
MDL Number MFCD00003451
Synonym Barium phosphate, dibasic
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1511118

  SDS
alfa aesar
1511118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1511130

  SDS
alfa aesar
1511130
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 233.31
Physical Form Powder
Quantity 50g
Color White
DOT Information DOT Class: 6.1, Packaging Group: III
EINECS Number 233-159-1
Chemical Name or Material Barium hydrogen phosphate
Molecular Formula BaHPO4
MDL Number MFCD00003451
CAS 10048-98-3
Synonym Barium phosphate, dibasic
Health Hazard 1 H302-H332: Harmful if swallowed. Harmful if inhaled.
Safety and Handling

H302-H332: Harmful if swallowed. Harmful if inhaled.