Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium hexacyanoferrate(II)

Chemical Identifiers

Chemical Name or Material Barium hexacyanoferrate (II)
CAS 13821-06-2
Molecular Formula Ba2Fe(CN)6
Formula Weight 486.62g/mol
MDL Number MFCD00168098
UN Number UN1564
Synonym Barium ferrocyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1037618

 
alfa aesar™
A10376-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA10376A1

 
alfa aesar™
A10376-A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50g
Quantity 50g
Chemical Name or Material Barium hexacyanoferrate (II)
CAS 13821-06-2
Molecular Formula Ba2Fe(CN)6
Formula Weight 486.62g/mol
MDL Number MFCD00168098
UN Number UN1564
Synonym Barium ferrocyanide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P261-P304+P340-P311-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P304+P340-P311-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.