Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium fluoride, ultra dry, 99.998% (metals basis)

Chemical Identifiers

Chemical Name or Material Barium fluoride
CAS 7787-32-8
Name Note Ultra dry
Formula Weight 175.36
MDL Number MFCD00003450
UN Number UN1564
Merck Index 14,974
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3861009

 
alfa aesar™
3861009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3861018

 
alfa aesar™
3861018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.36
Packaging Ampule
Chemical Name or Material Barium fluoride
TSCA TSCA
Odor Odorless
Name Note Ultra dry
Physical Form Powder
Assay 99.998%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00003450
CAS 7787-32-8
UN Number UN1564
Merck Index 14,974
Boiling Point 2260°C
Color White
Melting Point 1353°C
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
Density 4.83g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
RTECS Number CQ9100000

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

  • Ampouled under argon