missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium fluoride, Puratronic™, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1064736

 View more versions of this product

Catalog No. AA1064736



Specifications

Specifications

Barium fluoride
(metals basis)
MFCD00003450
14,974
OYLGJCQECKOTOL-UHFFFAOYSA-L
barium(2+);difluoride
62670
Powder
Puratronic™
Odorless
2260°C
500g
7787-32-8
BaF2
UN1564
barium fluoride
[F-].[F-].[Ba+2]
175.324
175.36
99.99%
4.83g/mL
1.4741
1353°C

Chemical Identifiers

7787-32-8
175.324
OYLGJCQECKOTOL-UHFFFAOYSA-L
62670
[F-].[F-].[Ba+2]
BaF2
MFCD00003450
barium fluoride
barium(2+);difluoride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

TSCA : TSCA

Documents