Please login to your online account to display your discounted pricing

Barium fluoride, 99%, pure, Acros Organics


Quantity 250g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  216832500

 View more versions of this product

Catalog No. AC216832500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Powder
Formula Weight 175.34
Quantity 250g
Color White to off-white
Merck Index 15, 970
EINECS Number 232-108-0
TSCA TSCA
Purity Grade Pure
Density 4.8900g/mL
RTECS Number CQ9100000
Chemical Name or Material Barium fluoride
Name Note 99%
Packaging Plastic bottle
Assay Percent Range 99%
Molecular Formula BaF2
Linear Formula BaF2
MDL Number MFCD00003450
CAS 7787-32-8
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Barium fluoride
Name Note 99%
CAS 7787-32-8
Purity Grade Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula BaF2
Linear Formula BaF2
Formula Weight 175.34
MDL Number MFCD00003450
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Toxic if swallowed.

SDS & Safety Documents