Please login to your online account to display your discounted pricing

Barium fluoride, 99%, pure, Acros Organics

$151.65 - $151.65

Chemical Identifiers

Chemical Name or Material Barium fluoride
Name Note 99%
CAS 7787-32-8
Purity Grade Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula BaF2
Linear Formula BaF2
Formula Weight 175.34
MDL Number MFCD00003450
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC216832500

  SDS
thermo fisher scientific
216832500
Plastic bottle 250g Each for $151.65
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Powder
Formula Weight 175.34
Quantity 250g
Color White to off-white
Merck Index 15, 970
Purity Grade Pure
TSCA TSCA
Density 4.8900g/mL
EINECS Number 232-108-0
RTECS Number CQ9100000
Packaging Plastic bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Toxic if swallowed.
Name Note 99%
Molecular Formula BaF2
CAS 7787-32-8
Chemical Name or Material Barium fluoride
Assay Percent Range 99%
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00003450
Linear Formula BaF2
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Toxic if swallowed.