Please login to your online account to display your discounted pricing

Barium Diphenylaminesulfonate, 0.1% (w/v) Aqueous Solution, Ricca Chemical

$51.24 - $51.24

Chemical Identifiers

Chemical Name or Material Barium Diphenylaminesulfonate
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 1300-92-1
CAS Min % 0.1
CAS Max % 0.1
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

884-4

 
ricca chemical company
RI8844
Poly natural 120mL Each for $51.24

R0884000500

 
ricca chemical company
RIR0884000500A
Plastic Bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear, colorless to light gray-blue
Quantity 120mL
Concentration or Composition Notes 0.1% (w/v)
Solvent or Matrix Aqueous Solution
CAS 7732-18-5
Chemical Name or Material Barium Diphenylaminesulfonate
CAS Max % 100.0
CAS Min % 0.1
CAS Min % 98.0
CAS Max % 0.1
Packaging Poly natural
CAS 1300-92-1