Please login to your online account to display your discounted pricing

Barium Diphenylamine Sulfonate, Reagent Grade, LabChem™

$484.80 - $484.80

Chemical Identifiers

Chemical Name or Material Barium Diphenylamine Sulfonate
CAS 6211-24-1
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C24H20N2S2O6Ba
Formula Weight 633.9
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC116258

 
lab chem
LC116258
25g Each for $484.80
Description & Specifications

Specifications

Formula Weight 633.9
Health Hazard 2 GHS H Statement
Harmful if swallowed or if inhaled.
Color White
Storage Information Room Temperature
Identification Passes Test
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
Chemical Name or Material Barium Diphenylamine Sulfonate
EINECS Number 228-278-0
Quantity 25g
CAS 6211-24-1
CAS Max % 100
Health Hazard 1 Warning
Packaging Amber Glass
Melting Point 300°C
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide; Barium; Nitrogen oxides
Solubility Soluble in water
Linear Formula C24H20N2S2O6Ba
Physical Form Powder
Purity Grade Reagent Grade
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed or if inhaled.