missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium Diphenylamine Sulfonate, For Chlorine, Chlorine Dioxide, Certified, 0.1% (w/v), LabChem™

$50.66 - $50.66

Chemical Identifiers

CAS 7732-18-5,6211-24-1
CAS Max % 99.9,0.1
Linear Formula C24H20N2S2O6Ba
Formula Weight 633.9
Chemical Name or Material Barium Diphenylamine Sulfonate, For Chlorine, Chlorine Dioxide
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
LC116307 Lab Chem
LC116307
Poly Bottle 125mL Each for $50.66
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,6211-24-1
C24H20N2S2O6Ba
Barium Diphenylamine Sulfonate, For Chlorine, Chlorine Dioxide
99.9,0.1
633.9
Certified Grade

Specifications

7732-18-5,6211-24-1
Passes Test
1g/mL
1g/mL
Clear, colorless
0.1% (w/v)
Soluble in water
Certified Grade
99.9,0.1
Poly Bottle
Liquid
C24H20N2S2O6Ba
633.9
125mL
Barium Diphenylamine Sulfonate, For Chlorine, Chlorine Dioxide

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature