missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium D-3-trifluoroacetylcamphorate

$40.24 - $40.24

Chemical Identifiers

CAS 36539-57-8
Molecular Formula C24H28BaF6O4
Molecular Weight (g/mol) 631.801
MDL Number MFCD00213545
InChI Key LGSBEDXATSQGDM-XITIRWOJSA-L
Synonym barium d-3-trifluoroacetylcamphorate
PubChem CID 90471783
IUPAC Name barium(2+);(1Z)-2,2,2-trifluoro-1-[(1S,4R)-4,7,7-trimethyl-3-oxo-2-bicyclo[2.2.1]heptanylidene]ethanolate
SMILES CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)[O-])C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)[O-])C)C.[Ba+2]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL00420MD View Documents Alfa Aesar™
L00420MD
250mg Each for $40.24
 
Specifications

Chemical Identifiers

36539-57-8
631.801
LGSBEDXATSQGDM-XITIRWOJSA-L
90471783
CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)[O-])C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)[O-])C)C.[Ba+2]
C24H28BaF6O4
MFCD00213545
barium d-3-trifluoroacetylcamphorate
barium(2+);(1Z)-2,2,2-trifluoro-1-[(1S,4R)-4,7,7-trimethyl-3-oxo-2-bicyclo[2.2.1]heptanylidene]ethanolate

Specifications

286°C to 288°C
36539-57-8
MFCD00213545
barium d-3-trifluoroacetylcamphorate
CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)[O-])C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)[O-])C)C.[Ba+2]
631.801
631.82
250mg
C24H28BaF6O4
UN1564
LGSBEDXATSQGDM-XITIRWOJSA-L
barium(2+);(1Z)-2,2,2-trifluoro-1-[(1S,4R)-4,7,7-trimethyl-3-oxo-2-bicyclo[2.2.1]heptanylidene]ethanolate
90471783
Barium D-3-trifluoroacetylcamphorate
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.

GHS P Statement
P261-P304+P340-P311-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.