Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium cyclohexanebutyrate, AAS, Ba 28.9%

Chemical Identifiers

Chemical Name or Material Barium cyclohexanebutyrate
CAS 62669-65-2
Purity Grade AAS Grade
Molecular Formula C20H34BaO4
Formula Weight 475.84
MDL Number MFCD00036397
UN Number UN1564
Synonym Cyclohexanebutyric acid barium salt; 4-Cyclohexylbutyric acid barium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8725703

  SDS
alfa aesar™
8725703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA8725706

  SDS
alfa aesar™
8725706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 475.84
Molecular Formula C20H34BaO4
Melting Point ≈225°C (dec.)
Purity Grade AAS Grade
Concentration Ba 28.9%
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
Synonym Cyclohexanebutyric acid barium salt; 4-Cyclohexylbutyric acid barium salt
TSCA TSCA
Health Hazard 1 Warning
Physical Form Powder
Chemical Name or Material Barium cyclohexanebutyrate
MDL Number MFCD00036397
CAS 62669-65-2
Quantity 1g
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
UN Number UN1564
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.