Please login to your online account to display your discounted pricing

Barium Crystalline dendritic solid, 99.9% (Metals basis), Alfa Aesar™

$354.09 - $354.09

Chemical Identifiers

Chemical Name or Material Barium
CAS 7440-39-3
Physical Form Crystalline Dendritic Solid
Assay Percent Range 99.9% (Metals basis)
MDL Number MFCD00134031
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1445414

  SDS
alfa aesar
1445414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1445406

  SDS
alfa aesar
1445406
5g Each for $354.09
Description & Specifications

Specifications

Physical Form Crystalline Dendritic Solid
Quantity 25g
Color Silver-gray
DOT Information DOT Class: 4.3, Packaging Group: II
EINECS Number 231-149-1
Health Hazard 1 H228-H261; Flammable solid. In contact with water releases flammable gas.
Odor Odorless
Appearance Silver Gray solid
Chemical Name or Material Barium
MDL Number MFCD00134031
Assay Percent Range 99.9% (Metals basis)
CAS 7440-39-3
Safety and Handling

H228-H261; Flammable solid. In contact with water releases flammable gas.