Please login to your online account to display your discounted pricing

Barium chloride, White beads, 99.99% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Barium chloride
CAS 10361-37-2
Physical Form Beads
Assay Percent Range 99.99% (Metals basis)
Molecular Formula BaCl2
Formula Weight 208.25
MDL Number MFCD00003445
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1368606

  SDS
alfa aesar
1368606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1368614

  SDS
alfa aesar
1368614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.25
Physical Form Beads
Quantity 5g
Color White
DOT Information DOT Class: 6.1, Packaging Group: III
Assay Percent Range 99.99% (Metals basis)
EINECS Number 233-788-1
CAS 10361-37-2
MDL Number MFCD00003445
Molecular Formula BaCl2
Chemical Name or Material Barium chloride
Health Hazard 1 H301-H331: Toxic if swallowed. Toxic if inhaled.
Odor Odorless
Safety and Handling

H301-H331: Toxic if swallowed. Toxic if inhaled.