missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium chloride, White beads, 99.99% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1368614

 View more versions of this product

Catalog No. AA1368614



Specifications

Specifications

Barium chloride
99.99% (Metals basis)
MFCD00003445
WDIHJSXYQDMJHN-UHFFFAOYSA-L
barium(2+);dichloride
25204
208.25
Odorless
25g
10361-37-2
BaCl2
barium chloride, barium dichloride, barium chloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
[Cl-].[Cl-].[Ba+2]
208.227
CHEBI:63317
Beads
White

Chemical Identifiers

10361-37-2
208.227
WDIHJSXYQDMJHN-UHFFFAOYSA-L
25204
barium(2+);dichloride
BaCl2
MFCD00003445
barium chloride, barium dichloride, barium chloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
CHEBI:63317
[Cl-].[Cl-].[Ba+2]
Safety and Handling

Safety and Handling

H301-H331: Toxic if swallowed. Toxic if inhaled.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 233-788-1

Documents