missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium chloride, ultra dry, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 1454906

 View more versions of this product

Catalog No. AA1454906



Description

Description

  • Ampouled under argon
Specifications

Specifications

Barium chloride
10361-37-2
BaCl2
UN1564
barium chloride, barium dichloride, barium chloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
[Cl-].[Cl-].[Ba+2]
208.227
CHEBI:63317
Beads
3.9g/mL
Odorless
Hygroscopic
White
5g
Ultra dry
(metals basis)
MFCD00003445
14,971
WDIHJSXYQDMJHN-UHFFFAOYSA-L
barium(2+);dichloride
25204
208.25
99.95%
−10 Mesh
Ampule
1560°C
963°C

Chemical Identifiers

10361-37-2
208.227
WDIHJSXYQDMJHN-UHFFFAOYSA-L
25204
barium(2+);dichloride
BaCl2
MFCD00003445
barium chloride, barium dichloride, barium chloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
CHEBI:63317
[Cl-].[Cl-].[Ba+2]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P301+P310-P304+P340-P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

RTECSNumber : CQ8750000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Documents