Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium chloride, ultra dry, 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Barium chloride
CAS 10361-37-2
Name Note Ultra dry
Formula Weight 208.25
MDL Number MFCD00003445
UN Number UN1564
Merck Index 14,971
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1454906

  SDS
alfa aesar™
1454906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1454914

  SDS
alfa aesar™
1454914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.25
Packaging Ampule
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99.95%
Name Note Ultra dry
Boiling Point 1560°C
TSCA TSCA
Merck Index 14,971
RTECS Number CQ8750000
MDL Number MFCD00003445
Mesh Size −10 Mesh
Storage Note 1
  • Ampouled under argon
Quantity 5g
Chemical Name or Material Barium chloride
Physical Form Beads
Assay Percent Notes (metals basis)
CAS 10361-37-2
Density 3.9g/mL
Melting Point 963°C
Health Hazard 1 Danger
Color White
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
Health Hazard 2 GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
Odor Odorless
UN Number UN1564
Sensitivity Hygroscopic

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P304+P340-P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

  • Ampouled under argon