Please login to your online account to display your discounted pricing

Barium Chloride Solution (20% w/v), Fisher Chemical


Quantity 20L
Packaging PolyPac™

Manufacturer: fisher scientific company  SB82220

 View more versions of this product

Catalog No. SB822-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 20L
Assay 19.5 to 20.5%
Physical Form Liquid
Color Colorless
NIST Traceable to NIST.
Health Hazard 2 WARNING!
MDL Number MFCD00003445
ChemAlert Storage Symbol Gray
Packaging PolyPac™
Identification Pass Test
Assay Percent Range 19.5 to 20.5 %
CAS 7732-18-5
Health Hazard 3 Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0
Name Note 20% w/v
CAS 10361-37-2
Chemical Name or Material Barium Chloride Solution

Specifications

Chemical Name or Material Barium Chloride Solution
Name Note 20% w/v
CAS 10361-37-2
CAS 7732-18-5
Assay 19.5 to 20.5%
Assay Percent Range 19.5 to 20.5 %
MDL Number MFCD00003445
Accessories
 

Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

SDS & Safety Documents