Please login to your online account to display your discounted pricing

Barium Chloride Solution (20% w/v), Fisher Chemical

$311.77 - $1,640.92

Chemical Identifiers

Chemical Name or Material Barium Chloride Solution
Name Note 20% w/v
CAS 10361-37-2
CAS 7732-18-5
Assay 19.5 to 20.5%
Assay Percent Range 19.5 to 20.5 %
MDL Number MFCD00003445
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB822-1

  SDS
fisher scientific company
SB8221
Poly Bottle 1L Each for $311.77
Case of 6 Each for $1,870.62

SB822-20

  SDS
fisher scientific company
SB82220
PolyPac™ 20L Each for $1,640.92

SB822-4

  SDS
fisher scientific company
SB8224
Poly Bottle 4L Each for $360.69
Case of 4 Each for $1,442.76
Description & Specifications

Specifications

Quantity 1L
Physical Form Liquid
Assay 19.5 to 20.5%
Color Colorless
Viscosity [NULL]
NIST Traceable to NIST.
Packaging Poly Bottle
Assay Percent Range 19.5 to 20.5 %
Identification Pass Test
MDL Number MFCD00003445
CAS 10361-37-2
ChemAlert Storage Symbol Gray
Health Hazard 2 WARNING!
CAS 7732-18-5
Health Hazard 3 Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0
Chemical Name or Material Barium Chloride Solution
Name Note 20% w/v
Safety and Handling
 

Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!