missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium chloride dihydrate, Puratronic, 99.999% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1064636

 View more versions of this product

Catalog No. AA1064636



Specifications

Specifications

Barium chloride dihydrate
10326-27-9
BaCl2H4O2
White powder
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.
O.O.[Cl-].[Cl-].[Ba+2]
244.257
CHEBI:86153
Powder
Odorless
113°C-2H2O
dihydrate
99.99% (Metal basis)
MFCD00149154
acmc-20alrc, bacl2.2h2o, barium 2+ dichloride dihydrate, barium 2+ dihydrate dichloride, barium chloride dihydrate, barium chloride, acs, dsstox_cid_131, dsstox_gsid_20131, dsstox_rid_75389, ksc174q7d
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+);dichloride;dihydrate
5284346
244.28 (208.25 Anhydrous)
3.097
White
500g

Chemical Identifiers

10326-27-9
244.257
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+);dichloride;dihydrate
BaCl2H4O2
MFCD00149154
acmc-20alrc, bacl2.2h2o, barium 2+ dichloride dihydrate, barium 2+ dihydrate dichloride, barium chloride dihydrate, barium chloride, acs, dsstox_cid_131, dsstox_gsid_20131, dsstox_rid_75389, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba+2]
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c

H301-H319-H332

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 600-412-6

RTECSNumber : CQ8751000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents