Please login to your online account to display your discounted pricing

Barium Chloride Dihydrate (Crystalline/Technical), Fisher Chemical


Quantity 50kg
Packaging Fiber Drum

Manufacturer: fisher scientific company  FLB3350

 View more versions of this product

Catalog No. B33-50

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 244.26
Melting Point 962°C
Purity Grade Technical Grade
Boiling Point 1560°C
Quantity 50kg
Density 3.86g/mL
UN Number 1564
DOT Information DOT Class 6.1, : Poison
Molecular Formula BaCl2 · 2 H2O
pH 5 to 8
Physical Form Solid
Decomposition Information Decomposition Information: > 100°C
Color White
Identification Pass Test
Health Hazard 3 Emergency Overview
Toxic if swallowed. Harmful by inhalation. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects.
NFPA
Health: 3
Flammability: 1
Instability: 0
Health Hazard 1 SIGNAL WORD: DANGER!
ChemAlert Storage Symbol Blue
Chemical Name or Material Barium Chloride Dihydrate
CAS 10326-27-9
Packaging Fiber Drum
MDL Number MFCD00149154
Health Hazard 2 DANGER!

Specifications

Chemical Name or Material Barium Chloride Dihydrate
CAS 10326-27-9
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula BaCl2 · 2 H2O
Formula Weight 244.26
MDL Number MFCD00149154
Accessories
 

SIGNAL WORD: DANGER!

Emergency Overview
Toxic if swallowed. Harmful by inhalation. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects.
NFPA
Health: 3
Flammability: 1
Instability: 0

DANGER!

SDS & Safety Documents