Please login to your online account to display your discounted pricing

Barium Chloride Dihydrate (Crystalline/Technical), Fisher Chemical


Quantity 10kg
Packaging Poly Pail

Manufacturer: fisher scientific company  FLB3310

 View more versions of this product

Catalog No. B33-10

  • / Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 1560°C
Purity Grade Technical Grade
Melting Point 962°C
Quantity 10kg
Formula Weight 244.26
UN Number 1564
DOT Information DOT Class 6.1, : Poison
Physical Form Solid
Molecular Formula BaCl2 · 2 H2O
Decomposition Information Decomposition Information: > 100°C
pH 5 to 8
Color White
Health Hazard 2 DANGER!
Health Hazard 1 SIGNAL WORD: DANGER!
CAS 10326-27-9
MDL Number MFCD00149154
Identification Pass Test
Packaging Poly Pail
Chemical Name or Material Barium Chloride Dihydrate
Health Hazard 3 Emergency Overview
Toxic if swallowed. Harmful by inhalation. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects.
NFPA
Health: 3
Flammability: 1
Instability: 0
ChemAlert Storage Symbol Blue

Specifications

Chemical Name or Material Barium Chloride Dihydrate
CAS 10326-27-9
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula BaCl2 · 2 H2O
Formula Weight 244.26
MDL Number MFCD00149154
Accessories
 

SIGNAL WORD: DANGER!

Emergency Overview
Toxic if swallowed. Harmful by inhalation. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects.
NFPA
Health: 3
Flammability: 1
Instability: 0

DANGER!

SDS & Safety Documents