Please login to your online account to display your discounted pricing

Barium Chloride Dihydrate (Crystalline/Technical), Fisher Chemical

$916.64 - $5,450.14

Chemical Identifiers

Chemical Name or Material Barium Chloride Dihydrate
CAS 10326-27-9
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula BaCl2 · 2 H2O
Formula Weight 244.26
MDL Number MFCD00149154
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B33-50

  SDS
fisher scientific company
FLB3350
Fiber Drum 50kg Each for $5,450.14

B33-10

  SDS
fisher scientific company
FLB3310
Poly Pail 10kg Each for $916.64
Description & Specifications

Specifications

Purity Grade Technical Grade
Formula Weight 244.26
Boiling Point 1560°C
Melting Point 962°C
Quantity 50kg
UN Number 1564
Density 3.86g/mL
DOT Information DOT Class 6.1, : Poison
Physical Form Solid
Molecular Formula BaCl2 · 2 H2O
Viscosity [NULL]
Decomposition Information Decomposition Information: > 100°C
Color White
pH 5 to 8
Chemical Name or Material Barium Chloride Dihydrate
Identification Pass Test
ChemAlert Storage Symbol Blue
Health Hazard 1 SIGNAL WORD: DANGER!
Health Hazard 3 Emergency Overview
Toxic if swallowed. Harmful by inhalation. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects.
NFPA
Health: 3
Flammability: 1
Instability: 0
Packaging Fiber Drum
MDL Number MFCD00149154
CAS 10326-27-9
Health Hazard 2 DANGER!

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

SIGNAL WORD: DANGER!

Emergency Overview
Toxic if swallowed. Harmful by inhalation. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects.
NFPA
Health: 3
Flammability: 1
Instability: 0

DANGER!