Please login to your online account to display your discounted pricing

Barium Chloride, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$127.31 - $1,018.82

Chemical Identifiers

Chemical Name or Material Barium Chloride
CAS 10326-27-9
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula BaCl2·2H2O
Formula Weight 244.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002397

 
avantor performance
097004
Poly Bottle 125g Each for $127.31

02002400

 
avantor performance
097005
Poly Bottle 2.5kg Each for $1,018.82

02002402

 
avantor performance
097001
Poly Bottle 500g Each for $244.08
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 244.27
Physical Form Crystals
Molecular Formula BaCl2·2H2O
CAS 10326-27-9
Specific Gravity 1L = 3.86kg
Packaging Poly Bottle
Name Note Dihydrate
Quantity 125g
Chemical Name or Material Barium Chloride