missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium chloride dihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 423455000

 View more versions of this product

Catalog No. AC423455000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1560.0°C
962.0°C
White
Crystals
99% min. (Complexometry)
BaCl2·2H2O
15, 967
Solubility in water: soluble. Other solubilities: soluble in methanol, practically insoluble in ethanol, acetone and, ethyl acetate
O.O.[Cl-].[Cl-].[Ba++]
244.26
CHEBI:86153
≥99%
14.0% to 16.0% (150°C)
Barium chloride dihydrate
10326-27-9
5.2 to 8.2 (5% soln. at 25°C)
500 g
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+) dihydrate dichloride
5284346
244.28
ACS Reagent
Plastic bottle

Chemical Identifiers

10326-27-9
244.26
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+) dihydrate dichloride
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba++]
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear eye protection/face protection.
If eye irritation per

GHS Signal Word: Danger

RTECSNumber : CQ8751000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only