Learn More
Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals
$64.03 - $613.59
Chemical Identifiers
CAS | 10326-27-9 |
---|---|
Molecular Formula | BaCl2H4O2 |
Molecular Weight (g/mol) | 244.26 |
MDL Number | MFCD00149154 |
InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
Synonym | barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d |
PubChem CID | 5284346 |
ChEBI | CHEBI:86153 |
SMILES | O.O.[Cl-].[Cl-].[Ba++] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1290530
|
Thermo Scientific Chemicals
A1290530 |
250 g |
Each for $64.03
|
|
|||||
AAA129050B
|
Thermo Scientific Chemicals
A129050B |
1000 g |
Each for $168.84
|
|
|||||
AAA129050I
|
Thermo Scientific Chemicals
A129050I |
5000 g |
Each for $613.59
|
|
|||||
Description
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBarium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.
Solubility
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.
Notes
Stable and hygroscopic in nature.
Chemical Identifiers
10326-27-9 | |
244.26 | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
5284346 | |
O.O.[Cl-].[Cl-].[Ba++] |
BaCl2H4O2 | |
MFCD00149154 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
CHEBI:86153 |
Specifications
113°C (in 2H2O) | |
250 g | |
MFCD00149154 | |
14,971 | |
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate. | |
O.O.[Cl-].[Cl-].[Ba++] | |
244.26 | |
CHEBI:86153 | |
≥99% | |
3.097 |
10326-27-9 | |
BaCl2H4O2 | |
UN1564 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
barium(2+) dihydrate dichloride | |
5284346 | |
244.28 (208.25 Anhydrous) | |
Odorless | |
Barium chloride dihydrate |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c
H301-H319-H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber : 233-788-1
RTECSNumber : CQ8751000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only