missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride, Certified, 20.0% (w/v) ±0.5%, LabChem™
$62.77 - $290.47
Chemical Identifiers
| CAS | 10326-27-9 |
|---|---|
| Molecular Formula | BaCl2H4O2 |
| Molecular Weight (g/mol) | 244.26 |
| MDL Number | MFCD00149154 |
| InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
| Synonym | barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d |
| PubChem CID | 5284346 |
| ChEBI | CHEBI:86153 |
| IUPAC Name | barium(2+) dihydrate dichloride |
| SMILES | O.O.[Cl-].[Cl-].[Ba++] |
Chemical Identifiers
| 10326-27-9 | |
| 244.26 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| 5284346 | |
| barium(2+) dihydrate dichloride |
| BaCl2H4O2 | |
| MFCD00149154 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| CHEBI:86153 | |
| O.O.[Cl-].[Cl-].[Ba++] |
Specifications
| 10326-27-9,7732-18-5 | |
| 82.52,17.48 | |
| 500 mL | |
| BaCl2·2H2O | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| barium(2+) dihydrate dichloride | |
| 5284346 | |
| 244.26 | |
| Certified | |
| Poly Bottle | |
| Hydrogen chloride; Barium | |
| Barium Chloride |
| Colorless | |
| Liquid | |
| BaCl2H4O2 | |
| MFCD00149154 | |
| Soluble in water | |
| O.O.[Cl-].[Cl-].[Ba++] | |
| 244.26 | |
| CHEBI:86153 | |
| 20.0% ±0.5% (w/v) | |
| Passes Test | |
| 1.15g/mL | |
| 1.15g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
Warning
Recommended Storage : Room Temperature