missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, Certified, 20.0% (w/v) ±0.5%, LabChem™

Supplier:  LabChem LC116051

 View more versions of this product

Catalog No. LC116051


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Barium Chloride
Colorless
Liquid
BaCl2H4O2
MFCD00149154
Soluble in water
O.O.[Cl-].[Cl-].[Ba++]
244.26
CHEBI:86153
20.0% ±0.5% (w/v)
Passes Test
1.15g/mL
1.15g/mL
10326-27-9,7732-18-5
82.52,17.48
500 mL
BaCl2·2H2O
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+) dihydrate dichloride
5284346
244.26
Certified
Poly Bottle
Hydrogen chloride; Barium

Chemical Identifiers

10326-27-9
244.26
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+) dihydrate dichloride
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba++]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions