Please login to your online account to display your discounted pricing

Barium Chloride, Certified, 12.0% (w/v) ±0.5%, LabChem™

$67.60 - $67.60

Chemical Identifiers

Chemical Name or Material Barium Chloride
CAS 7732-18-5
CAS Max % 89.11
CAS 10326-27-9
CAS Max % 10.89
Purity Grade Certified Grade
Linear Formula BaCl2·2H2O
Formula Weight 244.26
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC116022

 
lab chem
LC116022
1L Each for $67.60
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 12.0% ± 0.5% (w/v)
Formula Weight 244.26
Viscosity 1.03cSt
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Solubility Soluble in water
CAS Max % 89.11
CAS 7732-18-5
Density 1.08g/mL
Specific Gravity 1.08g/mL
Decomposition Information Hydrogen chloride; Barium
Color Clear, colorless
CAS 10326-27-9
Linear Formula BaCl2·2H2O
Packaging Poly Bottle
Physical Form Liquid
Storage Information Room Temperature
Identification Passes Test
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 10.89
Traceability to NIST Traceable to NIST
Quantity 1L
Chemical Name or Material Barium Chloride
Safety and Handling

Warning

GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.