Please login to your online account to display your discounted pricing

Barium Chloride, Anhydrous, Powder, Purified, J.T.Baker™

$222.73 - $898.28

Chemical Identifiers

Chemical Name or Material Barium Chloride
CAS 10361-37-2
Name Note Anhydrous
Purity Grade Purified
Molecular Formula BaCl2
Formula Weight 208.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002404

 
avantor performance
098005
Poly Bottle 2.5kg Each for $898.28

02002405

 
avantor performance
098001
Poly Bottle 500g Each for $222.73
Description & Specifications

Specifications

Formula Weight 208.23
CAS 10361-37-2
Physical Form Powder
Quantity 2.5kg
Packaging Poly Bottle
Specific Gravity 1L = 3.86kg
Chemical Name or Material Barium Chloride
Purity Grade Purified
Name Note Anhydrous
Molecular Formula BaCl2