missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium chloranilate trihydrate, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC401665000

 View more versions of this product

Catalog No. AC401665000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Barium chloranilate trihydrate
Plastic bottle
Brown
32458-20-1
MFCD00150175
Solubility in water: soluble
C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.O.O.O.[Ba]
398.34
398.34
Authentic
(for sulfate determination) passes
500g
C6BaCl2O4·3H2O
barium chloranilate, reag
JFBLKYVZQCFPJE-UHFFFAOYSA-N
barium;2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;trihydrate
131877766
Powder

Chemical Identifiers

32458-20-1
398.34
JFBLKYVZQCFPJE-UHFFFAOYSA-N
131877766
C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.O.O.O.[Ba]
C6BaCl2O4·3H2O
MFCD00150175
barium chloranilate, reag
barium;2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;trihydrate
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 236-567-8

TSCA : TSCA

SDS Documents