missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium chloranilate trihydrate, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 401661000

 View more versions of this product

Catalog No. AC401661000



Description

Description

Specifications

Specifications

Barium chloranilate trihydrate
Plastic bottle
Brown
32458-20-1
MFCD00078327
Solubility in water: soluble
O.O.O.[Ba].OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O
400.35
398.34
Authentic
(for sulfate determination) passes
100g
C6H8BaCl2O7
barium chloranilate, reag
JFBLKYVZQCFPJE-UHFFFAOYSA-N
barium;2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;trihydrate
131877766
Powder

Chemical Identifiers

32458-20-1
400.35
JFBLKYVZQCFPJE-UHFFFAOYSA-N
131877766
O.O.O.[Ba].OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O
C6H8BaCl2O7
MFCD00078327
barium chloranilate, reag
barium;2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;trihydrate
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 236-567-8

TSCA : TSCA

SDS Documents