Please login to your online account to display your discounted pricing

Barium chloranilate trihydrate, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  401660250

 View more versions of this product

Catalog No. AC401660250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 398.34
Physical Form Powder
Quantity 25g
Color Brown
EINECS Number 236-567-8
TSCA TSCA
Packaging Plastic bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
CAS 32458-20-1
Chemical Name or Material Barium chloranilate trihydrate
Molecular Formula C6BaCl2O4·3H2O
MDL Number MFCD00150175
Suitability (for sulfate determination) passes
Synonym Chloranilic acid barium salt trihydrate
Infrared Spectrum Authentic
Health Hazard 1 Warning

Specifications

Chemical Name or Material Barium chloranilate trihydrate
CAS 32458-20-1
Molecular Formula C6BaCl2O4·3H2O
Formula Weight 398.34
MDL Number MFCD00150175
Synonym Chloranilic acid barium salt trihydrate
Accessories

Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.

SDS & Safety Documents