Please login to your online account to display your discounted pricing

Barium Carbonate (Powder/Certified ACS), Fisher Chemical


Quantity 500g
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLB30500

 View more versions of this product

Catalog No. B30-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 1450°C
Melting Point 811°C
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Formula Weight 197.34
Quantity 500g
DOT Information DOT Class 6.1, : Poison
Assay 99.0 to 101.0%
Viscosity [NULL]
Color White
Density 4.430g/cm³
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Oxidizing Substances 0.005% max.
Sulfide 0.001% max.
CAS 513-77-9
ChemAlert Storage Symbol Blue
Titratable Base 0.002mEq/g max.
Additional Information Vapor Pressure: Negligible
Health Hazard 3 Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:0
Instability:0
Chloride 0.002% max.
Chemical Name or Material Barium Carbonate
Insoluble in Hydrochloric Acid 0.015% max.
Iron (Fe) 0.002% max.
Packaging Glass Bottle
Potassium (K) 0.005% max.
Assay Percent Range 99.0 to 101.0 %
Sodium (Na) 0.02% max.
Molecular Formula BaCO3
Strontium (Sr) 0.7% max.
MDL Number MFCD00003448
Synonym Carbonic Acid Barium Salt
Health Hazard 2 WARNING!
Calcium (Ca) 0.05% max.

Specifications

Chemical Name or Material Barium Carbonate
CAS 513-77-9
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Assay 99.0 to 101.0%
Assay Percent Range 99.0 to 101.0 %
Molecular Formula BaCO3
Formula Weight 197.34
MDL Number MFCD00003448
Synonym Carbonic Acid Barium Salt
Accessories
 

Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

SDS & Safety Documents