Please login to your online account to display your discounted pricing

Barium Carbonate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$424.44 - $2,348.42

Chemical Identifiers

Chemical Name or Material Barium Carbonate
CAS 513-77-9
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula BaCO3
Formula Weight 197.34
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002395

 
avantor performance
095005
Poly Bottle 2.5kg Each for $2,348.42

02002396

 
avantor performance
095001
Wide Mouth Amber Glass Bottle 500g Each for $424.44
Description & Specifications

Specifications

Formula Weight 197.34
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Barium Carbonate
Molecular Formula BaCO3
Physical Form Powder
Quantity 2.5kg
CAS 513-77-9
Specific Gravity 1L = 4.30kg
Packaging Poly Bottle