Please login to your online account to display your discounted pricing

Barium Carbonate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  095001

 View more versions of this product

Catalog No. 02002396

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 197.34
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Barium Carbonate
Molecular Formula BaCO3
Physical Form Powder
Quantity 500g
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 4.30kg
CAS 513-77-9

Specifications

Chemical Name or Material Barium Carbonate
CAS 513-77-9
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula BaCO3
Formula Weight 197.34