missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium carbonate, 99+%, for analysis, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 217495000

 View more versions of this product

Catalog No. AC217495000


 

Specifications Description & Specifications

Specifications

Barium carbonate
513-77-9
Powder
99+%
BaCO3
MFCD00003448
20ppm max.
10ppm max.
0.015% max. (in HCl)
Plastic bottle
10ppm max.
White
p.a.
For analysis
≥99.0% (BaCO3)
BaCO3
197.35
15, 965
4.4300g/mL
0.02% max.
10ppm max.
0.2% max.
5000ppm max.
500g

Chemical Identifiers

513-77-9
BaCO3
197.35
Barium carbonate, p.a.
For analysis
99+%
BaCO3
MFCD00003448
≥99.0% (BaCO3)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 208-167-3

RTECSNumber : CQ8600000

TSCA : TSCA