missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium carbonate, 99.8%, Alfa Aesar™

Manufacturer:  Alfa Aesar 14341A7

 View more versions of this product

Catalog No. AA14341A7



Specifications

Specifications

Barium carbonate
99.8%
MFCD00003448
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium(2+);carbonate
10563
1 Micron Powder
513-77-9
CBaO3
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
C(=O)([O-])[O-].[Ba+2]
197.335
197.35
5kg

Chemical Identifiers

513-77-9
197.335
AYJRCSIUFZENHW-UHFFFAOYSA-L
10563
C(=O)([O-])[O-].[Ba+2]
CBaO3
MFCD00003448
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
barium(2+);carbonate
Safety and Handling

Safety and Handling

EINECSNumber : 208-167-3

Documents