Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium calcium copper oxide (2-1-2), 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Barium calcium copper oxide (2-1-2)
MDL Number MFCD00211652
UN Number UN1564
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3919209

  SDS
alfa aesar™
3919209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3919218

  SDS
alfa aesar™
3919218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay Percent Notes (metals basis)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
Chemical Name or Material Barium calcium copper oxide (2-1-2)
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
UN Number UN1564
Assay 99.9%
Physical Form Fine Agglomerate & Ballmilled Powder
Quantity 10g
Health Hazard 1 Warning
MDL Number MFCD00211652
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.