Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium bromide, ultra dry, 99.998% (metals basis)

Chemical Identifiers

Chemical Name or Material Barium bromide
CAS 10553-31-8
Name Note Ultra dry
Formula Weight 297.14
MDL Number MFCD00003444
UN Number UN1564
Merck Index 14,968
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3567803

  SDS
alfa aesar™
3567803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3567806

  SDS
alfa aesar™
3567806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3567814

  SDS
alfa aesar™
3567814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 297.14
Packaging Ampule
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
Odor Odorless
Quantity 1g
UN Number UN1564
Melting Point 850°C
MDL Number MFCD00003444
Merck Index 14,968
Name Note Ultra dry
Health Hazard 1 Warning
Storage Note 1
  • Ampouled under argon
Density 4.781g/mL
Assay 99.998%
Color White
Assay Percent Notes (metals basis)
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Mesh Size −10 Mesh
Physical Form Beads
Sensitivity Hygroscopic
CAS 10553-31-8
TSCA TSCA
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Chemical Name or Material Barium bromide

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

  • Ampouled under argon