Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium bromide dihydrate, Puratronic™, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Barium bromide dihydrate
CAS 7791-28-8
Purity Grade Puratronic™
Formula Weight 333.17 (297.14 Anhydrous)
MDL Number MFCD00149153
UN Number UN1564
Merck Index 14,968
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1112506

  SDS
alfa aesar™
1112506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1112514

  SDS
alfa aesar™
1112514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1112522

  SDS
alfa aesar™
1112522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Barium bromide dihydrate
CAS 7791-28-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
TSCA TSCA
Purity Grade Puratronic™
Physical Form Powder
Assay 99.999%
Assay Percent Notes (metals basis)
Formula Weight 333.17 (297.14 Anhydrous)
MDL Number MFCD00149153
UN Number UN1564
Merck Index 14,968
Solubility Information Very soluble in water, soluble in methanol
Density 3.58g/mL
Odor Odorless
Color White
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.