Learn More
Barium bromide dihydrate, 99.3%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01219722
Description
Barium bromide dihydrate is used to purify radium and make zinc bromide. It also used  precursor to chemicals used in photograph.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Barium bromide dihydrate | |
Crystalline | |
99.30% | |
MFCD00149153 | |
HQQMRSGBIPFNSN-UHFFFAOYSA-L | |
barium(2+) dihydrate dibromide | |
333.17 (297.14 Anhydrous) |
7791-28-8 | |
100 g | |
BaBr2H4O2 | |
Very soluble in water,soluble in methanol | |
O.O.[Br-].[Br-].[Ba++] | |
333.17 | |
3.58 |
Chemical Identifiers
7791-28-8 | |
333.17 | |
HQQMRSGBIPFNSN-UHFFFAOYSA-L | |
O.O.[Br-].[Br-].[Ba++] |
BaBr2H4O2 | |
MFCD00149153 | |
barium(2+) dihydrate dibromide |
Safety and Handling
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 234-140-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only