Learn More
Barium bromide, anhydrous, 99%, Thermo Scientific Chemicals
$153.95 - $688.47
Description
Barium bromide, anhydrous is used as the chemical intermediate, and the spice cosmetics chemical additive. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10553-31-8 | |
| MFCD00003444 |
Specifications
| 850°C | |
| 10553-31-8 | |
| 50 g | |
| UN1564 | |
| Soluble in water | |
| 99% | |
| Hygroscopic | |
| Barium bromide, Anhydrous |
| White | |
| Powder | |
| MFCD00003444 | |
| 14,968 | |
| 297.14 | |
| Odorless | |
| 4.781 g/mL |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber : 234-140-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only