Learn More
Barium aluminum oxide, tech., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02264236
Description
Barium aluminum oxide is used for getter alloys in vacuum tubes. It also finds application as deoxidizer for copper, Fraryâs metal, lubricant for anode rotors in x-ray tubes and spark-plug alloys. It is used to prepare heat-resistant refractory and super-refractory concretes. Further, it gives a better protection against gamma radiation and X-rays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Barium aluminum oxide | |
Powder | |
MFCD00049716 | |
Barium aluminate | |
561.98 | |
Odorless |
12004-04-5 | |
500 g | |
UN1564 | |
Insoluble in water. | |
Technical |
Chemical Identifiers
12004-04-5 | |
MFCD00049716 | |
Barium aluminate |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber : 234-445-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only