Please login to your online account to display your discounted pricing

Barium Acetate, Reagent ACS, +99%, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC423430025

 View more versions of this product

Catalog No. AC423430025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Crystals
Formula Weight 255.43
Quantity 2.5kg
Color White
Assay 99.0 to 102.0%
EINECS Number 208-849-0
TSCA TSCA
Density 2.4600g/mL
RTECS Number AF4550000
Merck Index 15, 961
Chemical Name or Material Barium acetate
Heavy Metals (as Pb) 5ppm max.
Insoluble Matter 0.01% max.
Iron (Fe) 10ppm max.
Oxidizing Substances 0.005% max.
Packaging Plastic bottle
Potassium (K) 30ppm max.
Assay Percent Range 99+%
Sodium (Na) 50ppm max.
Molecular Formula C4H6O4Ba
Linear Formula (CH3CO2)2Ba
MDL Number MFCD00012447
CAS 543-80-6
Synonym Acetic acid, barium salt
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Calcium (Ca) 500ppm max.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Trace Analysis 2000ppm max.
Health Hazard 1 Warning
Chlorine (Cl) 10ppm max.

Specifications

Chemical Name or Material Barium acetate
CAS 543-80-6
Purity Grade ACS Reagent Grade
Assay 99.0 to 102.0%
Assay Percent Range 99+%
Molecular Formula C4H6O4Ba
Linear Formula (CH3CO2)2Ba
Formula Weight 255.43
MDL Number MFCD00012447
Synonym Acetic acid, barium salt
Accessories

Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents