Please login to your online account to display your discounted pricing

Barium Acetate (Crystalline/Certified ACS), Fisher Chemical

$1,054.69 - $1,054.69

Chemical Identifiers

Chemical Name or Material Barium Acetate
CAS 543-80-6
Purity Grade Certified ACS Grade
Physical Form Solid
Assay 99.0 to 102.0%
Assay Percent Range 99.0 to 102.0 %
Molecular Formula C4H6O4Ba
Linear Formula Ba(C2H3O2)2
Formula Weight 255.42
MDL Number MFCD00012447
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B2-421-2

  SDS
fisher scientific company
B24212
Poly Pail 2.5kg Each for $1,054.69
Description & Specifications

Specifications

Purity Grade Certified ACS Grade
Formula Weight 255.42
Quantity 2.5kg
DOT Information DOT Class 6.1, : Poison
Chemical Name or Material Barium Acetate
Identification Pass Test
Iron (Fe) 0.001% max.
Oxidizing Substances 0.005% max.
Packaging Poly Pail
Potassium (K) 0.003% max.
Physical Form Solid
Assay 99.0 to 102.0%
Assay Percent Range 99.0 to 102.0 %
Sodium (Na) 0.005% max.
Strontium (Sr) 0.2% max.
Linear Formula Ba(C2H3O2)2
ChemAlert Storage Symbol Blue
Synonym Acetic Acid Barium Salt
Color White
Calcium (Ca) 0.05% max.
Chloride 0.001% max.
Density 2.460g/cm³
Heavy Metals (as Pb) 5ppm max.
Insoluble Matter 0.01% max.
Molecular Formula C4H6O4Ba
MDL Number MFCD00012447
CAS 543-80-6