Please login to your online account to display your discounted pricing

Barium Acetate (Crystalline/Certified ACS), Fisher Chemical


Quantity 2.5kg
Packaging Poly Pail

Manufacturer: fisher scientific company  B24212

 View more versions of this product

Catalog No. B2-421-2

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified ACS Grade
Formula Weight 255.42
Quantity 2.5kg
DOT Information DOT Class 6.1, : Poison
Density 2.460g/cm³
Physical Form Solid
Color White
Assay 99.0 to 102.0%
Strontium (Sr) 0.2% max.
Packaging Poly Pail
Linear Formula Ba(C2H3O2)2
Assay Percent Range 99.0 to 102.0 %
Chloride 0.001% max.
Sodium (Na) 0.005% max.
Potassium (K) 0.003% max.
Identification Pass Test
Calcium (Ca) 0.05% max.
Iron (Fe) 0.001% max.
Chemical Name or Material Barium Acetate
Oxidizing Substances 0.005% max.
Synonym Acetic Acid Barium Salt
ChemAlert Storage Symbol Blue
Molecular Formula C4H6O4Ba
CAS 543-80-6
MDL Number MFCD00012447
Insoluble Matter 0.01% max.
Heavy Metals (as Pb) 5ppm max.

Specifications

Chemical Name or Material Barium Acetate
CAS 543-80-6
Purity Grade Certified ACS Grade
Physical Form Solid
Assay 99.0 to 102.0%
Assay Percent Range 99.0 to 102.0 %
Molecular Formula C4H6O4Ba
Linear Formula Ba(C2H3O2)2
Formula Weight 255.42
MDL Number MFCD00012447
Synonym Acetic Acid Barium Salt
Accessories
 

SDS & Safety Documents