Please login to your online account to display your discounted pricing

Barium Acetate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$205.78 - $822.69

Chemical Identifiers

Chemical Name or Material Barium Acetate
CAS 543-80-6
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (CH3COO)2Ba
Formula Weight 255.43
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002392

 
avantor performance
094205
Poly Bottle 2.5kg Each for $822.69

02002393

 
avantor performance
094201
Poly Bottle 500g Each for $205.78
Description & Specifications

Specifications

Formula Weight 255.43
Physical Form Crystals
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Barium Acetate
CAS 543-80-6
Molecular Formula (CH3COO)2Ba
Packaging Poly Bottle
Quantity 2.5kg
Specific Gravity 1L = 2.47kg