Please login to your online account to display your discounted pricing

Barium Acetate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  094201

 View more versions of this product

Catalog No. 02002393

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 255.43
Chemical Name or Material Barium Acetate
CAS 543-80-6
Packaging Poly Bottle
Specific Gravity 1L = 2.47kg
Molecular Formula (CH3COO)2Ba
Quantity 500g

Specifications

Chemical Name or Material Barium Acetate
CAS 543-80-6
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (CH3COO)2Ba
Formula Weight 255.43